1-Iodo-2,4-dinitrobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23659-06
B23659-14
CAAAB23659-06EA
128.61
CAD
CAAAB23659-06
CAAAB23659-14
1-Iodo-2,4-dinitrobenzene 98%
1-Iodo-2,4-dinitrobenzene
Purity: 98%
CAS No.: 709-49-9
Molecular Formula: C6H3IN2O4
Formula Weight: 294.01
Melting Point: 87-90°
Beilstein Reference: 1113584
Formula:
C₆H₃IN₂O₄ Melting Pt: 87…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039738 CAS Number: 709-49-9 EINECS: 211-908-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...