3-Nitrosalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

B23658-03 B23658-06
CAAAB23658-03EA 183.28 CAD
CAAAB23658-03 CAAAB23658-06
3-Nitrosalicylaldehyde 98%
3-Nitrosalicylaldehyde

Purity: 98% CAS No.: 5274-70-4 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Melting Point: 109-110° MDL No.: MFCD00041874

Formula: C₇H₅NO₄
Melting Pt: 109…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00041874
CAS Number: 5274-70-4
EINECS: 226-098-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR