3-Thiopheneboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Thienylboronic acid, Thiophene-3-boronic acid
B23637-03
B23637-06
B23637-14
CAAAB23637-03EA
52.93
CAD
CAAAB23637-03
CAAAB23637-06
CAAAB23637-14
3-Thiopheneboronic acid 98%
3-Thiopheneboronic acid
Purity: 98%
CAS No.: 6165-69-1
Molecular Formula: C4H5BO2S
Formula Weight: 127.96
Storage Temperature: -30°C to -10°C
Melting Point: 164-169°
Beilstein Reference: 113573
MDL No.: MFCD00151851
Formula:
C₄H₅BO₂S MW: 127.96 g/mol Melting Pt: 164…169 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00151851 CAS Number: 6165-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...