1-Iodo-4-(trifluoromethoxy)benzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23628-03
B23628-06
B23628-14
CAAAB23628-03EA
20.86
CAD
CAAAB23628-03
CAAAB23628-06
CAAAB23628-14
1-Iodo-4-(trifluoromethoxy)benzene 98%
1-Iodo-4-(trifluoromethoxy)benzene
Purity: 98%
CAS No.: 103962-05-6
Molecular Formula: C7H4F3IO
Formula Weight: 288.01
Flash Point: 77°(170°F)
Refractive Index: 1.5033
Boiling Point: 72-74°/15mm
Beilstein Reference: 6193794
MDL No.: MFCD00042411
Formula:
C₇H₄F₃IO Boiling Pt: 72…74 °C/15 mm Density: 1.909 Flash Pt: 77 °C (170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042411 CAS Number: 103962-05-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...