3-Cyclohexylpropionic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23613-18
B23613-30
CAAAB23613-18EA
103.33
CAD
CAAAB23613-18
CAAAB23613-30
3-Cyclohexylpropionic acid 98+%
3-Cyclohexylpropionic acid
Purity: 98+%
CAS No.: 701-97-3
Molecular Formula: C9H16O2
Formula Weight: 156.23
Flash Point: 130°(266°F)
Refractive Index: 1.4636
Odor: Unpleasant
Melting Point: 14-17°
Boiling Point: 276°
Beilstein Reference: 2042799
MDL No.: MFCD00001527
Formula:
C₉H₁₆O₂ Boiling Pt: 276 °C Melting Pt: 14…17 °C Density: 0.99 Flash Pt: 130 °C(266 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001527 CAS Number: 701-97-3 EINECS: 211-861-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...