2-(Methylamino)pyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23589-14
B23589-03
B23589-06
CAAAAB23589-14EA
306.52
CAD
CAAAAB23589-14
CAAAB23589-03
CAAAB23589-06
2-(Methylamino)pyridine 98%
2-(Methylamino)pyridine
Purity: 98%
CAS No.: 4597-87-9
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 87°(188°F)
Refractive Index: 1.5780
Melting Point: 14-15°
Boiling Point: 200-201°
Beilstein Reference: 108188
UN No.: 2810
MDL No.: MFCD00006250
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 200…201 °C Melting Pt: 14…15 °C Density: 1.060 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006250 CAS Number: 4597-87-9 EINECS: 224-997-9 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...