α-Bromo-2-chloro-4-fluorotoluene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(Bromomethyl)-2-chloro-4-fluorobenzene, 2-Chloro-4-fluorobenzyl bromide
B23565-06
B23565-14
CAAAB23565-06EA
29.39
CAD
CAAAB23565-06
CAAAB23565-14
α-Bromo-2-chloro-4-fluorotoluene 97%
α-Bromo-2-chloro-4-fluorotoluene
Purity: 97%
CAS No.: 45767-66-6
Molecular Formula: C7H5BrClF
Formula Weight: 223.48
Melting Point: 35-39°
Beilstein Reference: 3539265
UN No.: 3261
MDL No.: MFCD00236025
Formula:
C₇H₅BrClF MW: 223.47 g/mol Melting Pt: 35…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236025 CAS Number: 45767-66-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...