(3-Hydroxyphenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Hydroxyphenylacetic acid

B23563-03 B23563-06 B23563-14
CAAAB23563-03EA 34.44 CAD
CAAAB23563-03 CAAAB23563-06 CAAAB23563-14
(3-Hydroxyphenyl)acetic acid 99%
(3-Hydroxyphenyl)acetic acid

Purity: 99% CAS No.: 621-37-4 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 128-132° Beilstein Reference: 2086506

Formula: (C₆H₄(OH))CH₂COOH
MW: 152.15 g/mol
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 128…132 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004337
CAS Number: 621-37-4
EINECS: 210-684-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR