6-Amino-m-cresol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Amino-2-hydroxy-4-methylbenzene, 2-Hydroxy-4-methylaniline, Cetilistat Impurity 4, 4-Amino-3-hydroxytoluene, 2-Amino-5-methylphenol

B23560-09 B23560-18
CAAAB23560-09EA 71.42 CAD
CAAAB23560-09 CAAAB23560-18
6-Amino-m-cresol 98%
6-Amino-m-cresol

Purity: 98% CAS No.: 2835-98-5 Molecular Formula: C7H9NO Formula Weight: 123.16 Melting Point: 155-159° Beilstein Reference: 386144

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 259
Melting Pt: 162
Storage Temperature: Ambient
MDL Number: MFCD00007693
CAS Number: 2835-98-5
EINECS: 220-620-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR