2,4-Diaminotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Methyl-1,3-Phenylenediamine, 1,3-Diamino-4-methylbenzene, Toluene-2,4-diamine
B23495-0B
B23495-30
CAAAB23495-0BEA
252.8
CAD
CAAAB23495-0B
CAAAB23495-30
2,4-Diaminotoluene 98%
2,4-Diaminotoluene
Purity: 98%
CAS No.: 95-80-7
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point: 169°(336°F)
Odor: Recognizable
Boiling Point: 283-285°
Beilstein Reference: 2205839
UN No.: 1709
MDL No.: MFCD00007804
Formula:
C₇H₁₀N₂ Boiling Pt: 283…285 °C Melting Pt: 96…101 °C Density: 1.045 g/cm³ (20 °C) Flash Pt: 169 °C (336 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007804 CAS Number: 95-80-7 EINECS: 202-453-1 UN: 1709 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...