3-Acetylphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3'-Boronoacetophenone, 3-Acetylphenylboronic Acid, 3-Acetylbenzeneboronic acid, Dihydroxy(3-acetylphenyl)borane, (3-Acetylphenyl)metaboric acid, 3-Acetylphenylboronic acid, 3-Acetylbenzeneboronic acid
B23478-03
B23478-06
B23478-14
CAAAB23478-03EA
32.23
CAD
CAAAB23478-03
CAAAB23478-06
CAAAB23478-14
3-Acetylphenylboronic acid 97%
3-Acetylphenylboronic acid
Purity: 97%
CAS No.: 204841-19-0
Molecular Formula: C8H9BO3
Formula Weight: 163.97
Melting Point: ca 200-205°
Beilstein Reference: 8255673
Formula:
C₈H₉BO₃ Melting Pt: ∼ 200…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074678 CAS Number: 204841-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...