(±)-2-Phenyl-1-propanol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23473-18
B23473-30
CAAAB23473-18EA
100.65
CAD
CAAAB23473-18
CAAAB23473-30
(±)-2-Phenyl-1-propanol 97%
(±)-2-Phenyl-1-propanol
Purity: 97%
CAS No.: 1123-85-9
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 108°(226°F)
Refractive Index: 1.5262
Melting Point: -37°
Boiling Point: 220°
Beilstein Reference: 1906760
MDL No.: MFCD00004736
Formula:
C₆H₅CH(CH₃)CH₂OH Boiling Pt: 220 °C Melting Pt: –37 °C Density: 0.975 Flash Pt: 108 °C(226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004736 CAS Number: 1123-85-9 EINECS: 214-379-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...