Benzyl acetate 99%

Supplier: Thermo Scientific Chemicals
Warning

B23472-0E B23472-36
CAAAB23472-0EEA 202.24 CAD
CAAAB23472-0E CAAAB23472-36
Benzyl acetate 99%
Benzyl acetate

Purity: 99% CAS No.: 140-11-4 Molecular Formula: CH3CO2CH2C6H5 Formula Weight: 150.18 Flash Point: 95°(203°F) Physical Form: Liquid Density: 1.053 Refractive Index: 1.5020 Appearance: Colorless Odor: Fruit-like Melting Point: -51° Boiling Point: 205-207° Beilstein Reference: 1908121 Merck Reference: 14,1123 MDL No.: MFCD00008712

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 205…207 °C
Melting Pt: –51 °C
Density: 1.053
Flash Pt: 95 °C(203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008712
CAS Number: 140-11-4
EINECS: 205-399-7
Merck Index: 14,1123

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR