Ethyl octanoate 99%
Supplier: Thermo Scientific Chemicals
Warning
|
B23450-22
B23450-36
CAAAB23450-22EA
42.28
CAD
CAAAB23450-22
CAAAB23450-36
Ethyl octanoate 99%
Ethyl octanoate
Purity: 99%
CAS No.: 106-32-1
Molecular Formula: C10H20O2
Formula Weight: 172.27
Flash Point: 79°(174°F)
Refractive Index: 1.4170
Odor: Ester-like
Melting Point: -47°
Boiling Point: 206-208°
Merck Reference: 14,3778
MDL No.: MFCD00009552
Formula:
C₁₀H₂₀O₂ Boiling Pt: 206…208 °C Melting Pt: –47 °C Density: 0.872 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009552 CAS Number: 106-32-1 EINECS: 203-385-5 Merck Index: 14,3778 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...