1-Bromo-2-ethylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

B23449-06 B23449-14 B23449-22
CAAAB23449-06EA 77.1 CAD
CAAAB23449-06 CAAAB23449-14 CAAAB23449-22
1-Bromo-2-ethylbenzene 98%
1-Bromo-2-ethylbenzene

Purity: 98% CAS No.: 1973-22-4 Molecular Formula: C8H9Br Formula Weight: 185.06 Flash Point: 71°(159°F) Refractive Index: 1.5485 Melting Point: -68° Boiling Point: 199-200° Beilstein Reference: 1929735 MDL No.: MFCD00000077

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 199…200 °C
Melting Pt: –68 °C
Density: 1.338
Flash Pt: 71 °C(159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000077
CAS Number: 1973-22-4
EINECS: 217-823-8
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR