D-Norvaline 99%
Supplier: Thermo Scientific Chemicals
B23444-03
B23444-06
B23444-14
CAAAB23444-03EA
42.03
CAD
CAAAB23444-03
CAAAB23444-06
CAAAB23444-14
D-Norvaline 99%
D-Norvaline
Purity: 99%
CAS No.: 2013-12-9
Molecular Formula: C5H11NO2
Formula Weight: 117.15
Melting Point:
Formula:
C₅H₁₁NO₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008097 CAS Number: 2013-12-9 EINECS: 217-936-2 Merck Index: 14,6716 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...