2-Amino-6-methylbenzothiazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Methylbenzothiazol-2-ylamine
B23422-14
B23422-22
CAAAB23422-14EA
76.79
CAD
CAAAB23422-14
CAAAB23422-22
2-Amino-6-methylbenzothiazole 99%
2-Amino-6-methylbenzothiazole
Purity: 99%
CAS No.: 2536-91-6
Molecular Formula: C8H8N2S
Formula Weight: 164.22
Melting Point: 139-142°
MDL No.: MFCD00005789
Formula:
C₈H₈N₂S MW: 164.23 g/mol Boiling Pt: 321 Melting Pt: 142 Storage Temperature: Ambient |
MDL Number:
MFCD00005789 CAS Number: 2536-91-6 EINECS: 219-801-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...