4-Amino-2-chlorophenol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloro-4-hydroxyaniline

B23420-03 B23420-06 B23420-14
CAAAB23420-03EA 86.27 CAD
CAAAB23420-03 CAAAB23420-06 CAAAB23420-14
4-Amino-2-chlorophenol 99%
4-Amino-2-chlorophenol

Purity: 99% CAS No.: 3964-52-1 Molecular Formula: C6H6ClNO Formula Weight: 143.57 Melting Point: 150-153° MDL No.: MFCD00128891

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 150…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00128891
CAS Number: 3964-52-1
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR