3-(Hydroxymethyl)benzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Benzenedimethanol, m-Xylene-α,α'-diol, m-Phenylenedimethanol

B23417-03 B23417-06 B23417-14
CAAAB23417-03EA 52.46 CAD
CAAAB23417-03 CAAAB23417-06 CAAAB23417-14
3-(Hydroxymethyl)benzyl alcohol 98%
3-(Hydroxymethyl)benzyl alcohol

Purity: 98% CAS No.: 626-18-6 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 56-60° Boiling Point: 154-159°/13mm MDL No.: MFCD00004649

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 283
Melting Pt: 58
Storage Temperature: Ambient
MDL Number: MFCD00004649
CAS Number: 626-18-6
EINECS: 210-934-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR