2,2'-Dimethoxybiphenyl 97%

Supplier: Thermo Scientific Chemicals

B23411-06 B23411-14 B23411-22
CAAAB23411-06EA 105.54 CAD
CAAAB23411-06 CAAAB23411-14 CAAAB23411-22
2,2'-Dimethoxybiphenyl 97%
2,2'-Dimethoxybiphenyl

Purity: 97% CAS No.: 4877-93-4 Molecular Formula: C14H14O2 Formula Weight: 214.26 Melting Point: 152-156° MDL No.: MFCD00067073

Formula: C₁₄H₁₄O₂
MW: 214.26
Boiling Pt: 307.5
Melting Pt: 152…156 °C
Density: 1.268
Storage Temperature: Ambient
MDL Number: MFCD00067073
CAS Number: 4877-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR