3-Amino-2,4,6-triiodobenzoic acid, (max. 4% H₂O) 99% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

B23405-06 B23405-14
CAAAB23405-06EA 74.26 CAD
CAAAB23405-06 CAAAB23405-14
3-Amino-2,4,6-triiodobenzoic acid, (max. 4% H₂O) 99% (dry weight)
3-Amino-2,4,6-triiodobenzoic acid

Purity: 99% CAS No.: 3119-15-1 Molecular Formula: C7H4I3NO2 Formula Weight: 514.83 Melting Point: ca 196° dec. MDL No.: MFCD00007681

Formula: C₇H₄I₃NO₂
Boiling Pt: 459 °C (1013 hPa)
Melting Pt: ∼ 196 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007681
CAS Number: 3119-15-1
EINECS: 221-493-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR