4-Chloro-2-nitrotoluene 99%

Supplier: Thermo Scientific Chemicals
Warning

B23382-22 B23382-36
CAAAB23382-22EA 57.2 CAD
CAAAB23382-22 CAAAB23382-36
4-Chloro-2-nitrotoluene 99%
4-Chloro-2-nitrotoluene

Purity: 99% CAS No.: 89-59-8 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Flash Point: 120°(248°F) Melting Point: 34-38° Boiling Point: 242° UN No.: 3457 MDL No.: MFCD00007215

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 242 °C
Melting Pt: 34…38 °C
Density: 1.2559 g/cm³ (20 °C)
Flash Pt: 120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007215
CAS Number: 89-59-8
EINECS: 201-921-2
UN: 3457
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR