2-Amino-6-methoxybenzothiazole 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23380-09
B23380-18
CAAAB23380-09EA
46.14
CAD
CAAAB23380-09
CAAAB23380-18
2-Amino-6-methoxybenzothiazole 98%
2-Amino-6-methoxybenzothiazole
Purity: 98%
CAS No.: 1747-60-0
Molecular Formula: C8H8N2OS
Formula Weight: 180.23
Odor: Odorless
Melting Point: 161-164°
MDL No.: MFCD00005787
Formula:
C₈H₈N₂OS Boiling Pt: 348 °C (1013 hPa) Melting Pt: 161…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005787 CAS Number: 1747-60-0 EINECS: 217-130-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...