2-Methoxybenzophenone 98%
Supplier: Thermo Scientific Chemicals
B23376-03
B23376-06
CAAAB23376-03EA
101.75
CAD
CAAAB23376-03
CAAAB23376-06
2-Methoxybenzophenone 98%
2-Methoxybenzophenone
Purity: 98%
CAS No.: 2553-04-0
Molecular Formula: C14H12O2
Formula Weight: 212.25
Melting Point: 36-38°
Boiling Point: 149-150°/1mm
MDL No.: MFCD00017163
Formula:
C₁₄H₁₂O₂ Boiling Pt: 149…150 °C/1 mm Melting Pt: 36…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017163 CAS Number: 2553-04-0 EINECS: 219-857-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...