Light green SF yellowish
Supplier: Thermo Scientific Chemicals
Synonyms:
Dihydrogen (ethyl)[4-[p-[ethyl(m-sulphonatobenzyl)amino]-p'-sulphonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulphonatobenzyl)ammonium disodium salt, Acid Green 5, Dihydrogen (ethyl)[4-[p-[ethyl(m-sulfonatobenzyl)amino]-p'-sulfonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulfonatobenzyl)ammonium disodium salt, Light Green 2G, S or 2GN, CI 42095, acid Green 5, Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulfonatobenzyl)amino)phenyl)(4-sulfonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulfonate, Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-sulphonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulphonate, Food Green n°2, Acid Green F, Fast acid Green N, acid Green F, CN DcL-30
Useful as a general plasma stain, as a histological stain, and a cytological counterstain
CAS No.: 5141-20-8
Molecular Formula: C37H34N2Na2O9S3
Formula Weight: 792.86
Melting Point: ca 288° dec.
Beilstein Reference: 5717828
Merck Reference: 14,5485
MDL No.: MFCD00012121
Formula:
C₃₇H₃₄N₂Na₂O₉S₃ MW: 792.86 g/mol Melting Pt: ca 288 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012121 CAS Number: 5141-20-8 EINECS: 225-906-5 Merck Index: 14,5485 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...