2-Ethoxynaphthalene 99%

Supplier: Thermo Scientific Chemicals

B23327-14 B23327-22 B23327-36
CAAAAB23327-14EA 30.97 CAD
CAAAAB23327-14 CAAAB23327-22 CAAAB23327-36
2-Ethoxynaphthalene 99%
2-Ethoxynaphthalene

Purity: 99% CAS No.: 93-18-5 Molecular Formula: C12H12O Formula Weight: 172.23 Flash Point: 134°(273°F) Melting Point: 35-37° Boiling Point: 282° Beilstein Reference: 2042397 Merck Reference: 14,3752 MDL No.: MFCD00016808

Formula: C₁₂H₁₂O
Boiling Pt: 282 °C
Melting Pt: 35…37 °C
Density: 1.064 g/cm³ (20 °C)
Flash Pt: 134 °C(273 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016808
CAS Number: 93-18-5
EINECS: 202-226-7
Merck Index: 14,3752

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR