2-Chlorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B23324-03 B23324-06 B23324-14
CAAAB23324-03EA 65.57 CAD
CAAAB23324-03 CAAAB23324-06 CAAAB23324-14
2-Chlorophenylboronic acid 97%
2-Chlorophenylboronic acid

Purity: 97% CAS No.: 3900-89-8 Molecular Formula: C6H6BClO2 Formula Weight: 156.38 Melting Point: ca 95-100° Beilstein Reference: 3030414

Formula: C₆H₆BClO₂
Melting Pt: ∼ 95…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00674012
CAS Number: 3900-89-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR