2,3-Dimethylquinoxaline 97%

Supplier: Thermo Scientific Chemicals
Danger

B23313-06 B23313-14
CAAAB23313-06EA 50.09 CAD
CAAAB23313-06 CAAAB23313-14 CAAAB23313-22
2,3-Dimethylquinoxaline 97%
2,3-Dimethylquinoxaline

Purity: 97% CAS No.: 2379-55-7 Molecular Formula: C10H10N2 Formula Weight: 158.20 Melting Point: 102-107° Beilstein Reference: 114832

Formula: C₁₀H₁₀N₂
Boiling Pt: 125 - 130 °C (1.5 -rr)
Melting Pt: 102…107 °C
Density: 1.226 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006728
CAS Number: 2379-55-7
EINECS: 219-162-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR