4-Allylanisole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Allyl-4-methoxybenzene, Estragole
B23311-14
B23311-22
CAAAB23311-14EA
57.2
CAD
CAAAB23311-14
CAAAB23311-22
4-Allylanisole 98%
4-Allylanisole
Purity: 98%
CAS No.: 140-67-0
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 81°(177°F)
Refractive Index: 1.5210
Boiling Point: 215-216°
Beilstein Reference: 1099454
Merck Reference: 14,3705
MDL No.: MFCD00008653
Formula:
C₁₀H₁₂O Boiling Pt: 215…216 °C Melting Pt: 22.5 °C Density: 0.965 Flash Pt: 81 °C(177 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008653 CAS Number: 140-67-0 EINECS: 205-427-8 Merck Index: 14,3705 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...