1-Iodo-3,3,3-trifluoropropane 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,1,1-Trifluoro-3-iodopropane, 3-Iodo-1,1,1-trifluoropropane
B23284-03
B23284-06
CAAAB23284-03EA
45.66
CAD
CAAAB23284-03
CAAAB23284-06
1-Iodo-3,3,3-trifluoropropane 98+%
1-Iodo-3,3,3-trifluoropropane
Purity: 98+%
CAS No.: 460-37-7
Molecular Formula: CF3CH2CH2I
Formula Weight: 223.96
Flash Point: None
Refractive Index: 1.4200
Boiling Point: 89-90°
Beilstein Reference: 1698182
MDL No.: MFCD00038531
Formula:
C₃H₄F₃I Boiling Pt: 89…90 °C Density: 1.911 Storage Temperature: Ambient |
MDL Number:
MFCD00038531 CAS Number: 460-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...