Methyl isonicotinate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23263-14
B23263-22
B23263-36
CAAAB23263-14EA
46.61
CAD
CAAAB23263-14
CAAAB23263-22
CAAAB23263-36
Methyl isonicotinate 98%
Methyl isonicotinate
Purity: 98%
CAS No.: 2459-09-8
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Flash Point: 82°(180°F)
Refractive Index: 1.5135
Odor: Characteristic
Melting Point: 8-8.5°
Boiling Point: 207-209°
MDL No.: MFCD00006427
Formula:
C₇H₇NO₂ Boiling Pt: 207…209 °C Melting Pt: 8…8.5 °C Density: 1.016 Flash Pt: 82 °C(180 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006427 CAS Number: 2459-09-8 EINECS: 219-456-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...