2-Ethylhexyl acetate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23257-AK
B23257-AU
CAAAB23257-AKEA
44.08
CAD
CAAAB23257-AK
CAAAB23257-AU
2-Ethylhexyl acetate 99%
2-Ethylhexyl acetate
Purity: 99%
CAS No.: 103-09-3
Molecular Formula: C10H20O2
Formula Weight: 172.27
Flash Point: 76°(169°F)
Refractive Index: 1.4200
Odor: Pleasant
Melting Point: -45°
Boiling Point: 199-200°
Beilstein Reference: 1758321
Merck Reference: 14,6763
MDL No.: MFCD00027249
Formula:
C₁₀H₂₀O₂ Boiling Pt: 199…200 °C Melting Pt: –45 °C Density: 0.870 Flash Pt: 76 °C(169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00027249 CAS Number: 103-09-3 EINECS: 203-079-1 Merck Index: 14,6763 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...