3-Brompropylamine hydrobromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23254-14
B23254-22
CAAAB23254-14EA
76.47
CAD
CAAAB23254-14
CAAAB23254-22
3-Brompropylamine hydrobromide 98%
Formula:
BrH₂CCH₂CH₂NH₃Br Melting Pt: 172…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012912 CAS Number: 5003-71-4 EINECS: 225-675-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...