N-Isopropylbenzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

B23243-22 B23243-36
CAAAB23243-22EA 88.01 CAD
CAAAB23243-22 CAAAB23243-36
N-Isopropylbenzylamine 97%
N-Isopropylbenzylamine

Purity: 97% CAS No.: 102-97-6 Molecular Formula: C10H15N Formula Weight: 149.24 Flash Point: 87°(190°F) Refractive Index: 1.5020 Boiling Point: 199-201° MDL No.: MFCD00008863

Formula: C₆H₅CH₂N(CH₃)(CH₂CH₃)
Boiling Pt: 199…201 °C
Melting Pt: ∼ 0 °C
Density: 0.910
Flash Pt: 87 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008863
CAS Number: 102-97-6
EINECS: 203-067-6
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR