4-Acetylphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Acetylphenyl)metaboric acid, 4-Acetylbenzeneboronic acid, Dihydroxy(4-acetylphenyl)borane

B23234-03 B23234-06 B23234-14
CAAAB23234-03EA 39.97 CAD
CAAAB23234-03 CAAAB23234-06 CAAAB23234-14
4-Acetylphenylboronic acid 98%
4-Acetylphenylboronic acid

Purity: 98% CAS No.: 149104-90-5 Molecular Formula: C8H9BO3 Formula Weight: 163.97 Melting Point: ca 240° dec. Beilstein Reference: 7869162

Formula: C₈H₉BO₃
Melting Pt: ∼ 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01074667
CAS Number: 149104-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR