3-Iodopyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

B23225-06 B23225-14
CAAAB23225-06EA 221.2 CAD
CAAAB23225-06 CAAAB23225-14
3-Iodopyridine 99%
3-Iodopyridine

Purity: 99% CAS No.: 1120-90-7 Molecular Formula: C5H4IN Formula Weight: 205.00 Flash Point: 106°(222°F) Melting Point: 53-56° Boiling Point: 90-92°/14mm Beilstein Reference: 106059 MDL No.: MFCD00023553

Formula: C₅H₄IN
MW: 205 g/mol
Boiling Pt: 90…92 °C/14 mm
Melting Pt: 53…56 °C
Flash Pt: 106 °C(222 °F)
Storage Temperature: Ambient
MDL Number: MFCD00023553
CAS Number: 1120-90-7
EINECS: 214-322-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR