Fluorene-9-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
9H-Fluorene-9-carboxylic acid
B23224-06
B23224-14
B23224-22
CAAAB23224-06EA
63.99
CAD
CAAAB23224-06
CAAAB23224-14
CAAAB23224-22
Fluorene-9-carboxylic acid 97%
Fluorene-9-carboxylic acid
Purity: 97%
CAS No.: 1989-33-9
Molecular Formula: C14H10O2
Formula Weight: 210.23
Flash Point: 230°(446°F)
Melting Point: 226-232°
Beilstein Reference: 1911728
MDL No.: MFCD00001136
Formula:
C₁₄H₁₀O₂ Boiling Pt: 365 °C (1013 hPa) Melting Pt: 226…232 °C Density: 1.333 g/cm³ (20 °C) Flash Pt: 230 °C(446 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001136 CAS Number: 1989-33-9 EINECS: 217-866-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...