2-Methylaminoethanol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Methyl-2-ethanolamin, N-Methyl-2-aminoethanol, 2-(Methylamino)ethanol

B23180-AE B23180-AP
CAAAB23180-AEEA 52.14 CAD
CAAAB23180-AE CAAAB23180-AP
2-Methylaminoethanol 99%
2-Methylaminoethanol

Purity: 99% CAS No.: 109-83-1 Molecular Formula: C5H7NO Formula Weight: 75.11 Flash Point: 74°(165°F) Refractive Index: 1.4400 Odor: Ammonia-like Melting Point: -5° Boiling Point: 159° Beilstein Reference: 1071196 Merck Reference: 14,6015 UN No.: 2735 MDL No.: MFCD00002839

Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 159 °C
Melting Pt: –5 °C
Density: 0.939
Flash Pt: 74 °C (165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002839
CAS Number: 109-83-1
EINECS: 203-710-0
UN: 2735
ADR: 8,III
Merck Index: 14,6015

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR