Tetraethylammonium fluoride hydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrylammonium fluoride hydrate, Tetraethylammonium fluoride xhydrate

B23173-06 B23173-14 B23173-22
CAAAB23173-06EA 133.04 CAD
CAAAB23173-06 CAAAB23173-14 CAAAB23173-22
Tetraethylammonium fluoride hydrate 97%
Tetraethylammonium fluoride hydrate

Purity: 97% CAS No.: 98330-04-2 Molecular Formula: (CH3CH2)4NF.xH2O Formula Weight: 149.26(anhy) Beilstein Reference: 4152659 MDL No.: MFCD00149990

Formula: C₈H₂₀FN·xH₂O
MW: 149.25 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149990
CAS Number: 98330-04-2
EINECS: 211-559-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR