3,4-Dimethylthiophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Dimethyl-4-mercaptobenzene, 4-Mercapto-o-xylene, 3,4-Dimethylbenzene-1-thiol, 3,4-Dimethylbenzenethiol

B23168-03 B23168-06 B23168-14
CAAAB23168-03EA 78.53 CAD
CAAAB23168-03 CAAAB23168-06 CAAAB23168-14
3,4-Dimethylthiophenol 98%
3,4-Dimethylthiophenol

Purity: 98% CAS No.: 18800-53-8 Molecular Formula: C8H10S Formula Weight: 138.23 Flash Point: 90°(194°F) Refractive Index: 1.5736 Odor: Unpleasant Boiling Point: 218° Beilstein Reference: 2040353 UN No.: 2810 MDL No.: MFCD00010023

Formula: C₈H₁₀S
MW: 138.23 g/mol
Boiling Pt: 218 °C
Melting Pt: 54…56 °C
Density: 1.027
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010023
CAS Number: 18800-53-8
EINECS: 242-587-8
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR