2,4-Dimethylthiophenol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4-Dimethylbenzene-1-thiol, 1,3-Dimethyl-4-mercaptobenzene, 4-Mercapto-m-xylene, 2,4-Dimethylbenzenethiol

B23161-06 B23161-14 B23161-22
CAAAB23161-06EA 116.92 CAD
CAAAB23161-06 CAAAB23161-14 CAAAB23161-22
2,4-Dimethylthiophenol 95%
2,4-Dimethylthiophenol

Purity: 95% CAS No.: 13616-82-5 Molecular Formula: C8H10S Formula Weight: 138.23 Flash Point: 87°(189°F) Refractive Index: 1.5690 Odor: Unpleasant Boiling Point: 207-208° UN No.: 2810 MDL No.: MFCD00010019

Formula: C₈H₁₀S
MW: 138.23 g/mol
Boiling Pt: 207…208 °C
Density: 1.020
Flash Pt: 87 °C (189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010019
CAS Number: 13616-82-5
EINECS: 237-100-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR