2-Tolylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23154-03
B23154-06
B23154-14
CAAAB23154-03EA
69.2
CAD
CAAAB23154-03
CAAAB23154-06
CAAAB23154-14
2-Tolylboronic acid 98%
2-Tolylboronic acid
Purity: 98%
CAS No.: 16419-60-6
Molecular Formula: C7H9BO2
Formula Weight: 135.96
Melting Point: ca 162-164°
Beilstein Reference: 2935796
Formula:
C₇H₉BO₂ Melting Pt: ∼ 162…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00093526 CAS Number: 16419-60-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...