Methyl-p-toluate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23111-22
B23111-36
CAAAB23111-22EA
58.14
CAD
CAAAB23111-22
CAAAB23111-36
Methyl-p-toluate 99%
Methyl p-toluate
Purity: 99%
CAS No.: 99-75-2
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 89°(192°F)
Odor: Stench
Boiling Point: 221-224°
Beilstein Reference: 1100609
MDL No.: MFCD00008441
Formula:
C₉H₁₀O₂ Boiling Pt: 221…224 °C Melting Pt: 32…35 °C Density: 1.060 Flash Pt: 89 °C(192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008441 CAS Number: 99-75-2 EINECS: 202-784-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...