2-Fluorophenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dihydroxy(2-fluorophenyl)borane, 2-Fluorobenzeneboronic acid, (2-Fluorophenyl)metaboric acid, (2-Fluorophenyl)boronic acid
B23103-03
B23103-06
CAAAB23103-03EA
70.15
CAD
CAAAB23103-03
CAAAB23103-06
CAAAB23103-14
2-Fluorophenylboronic acid 98%
2-Fluorophenylboronic acid
Purity: 98%
CAS No.: 1993-03-9
Molecular Formula: C6H6BFO2
Formula Weight: 139.92
Melting Point: 100-108°
Beilstein Reference: 3030413
Formula:
C₆H₆BFO₂ MW: 139.92 g/mol Melting Pt: 100…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00674013 CAS Number: 1993-03-9 |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...