Pyromellitic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

B23099-22 B23099-36
CAAAB23099-22EA 77.89 CAD
CAAAB23099-22 CAAAB23099-36
Pyromellitic acid 96%
Pyromellitic acid

Purity: 96% CAS No.: 89-05-4 Molecular Formula: C10H6O8 Formula Weight: 254.15 Melting Point: 281-285° Merck Reference: 14,8004 MDL No.: MFCD00002471

Formula: C₁₀H₆O₈
Boiling Pt: 516 °C (1013 hPa)
Melting Pt: 281…285 °C
Density: 1.79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002471
CAS Number: 89-05-4
EINECS: 201-879-5
Merck Index: 14,8004

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR