(Ethyl)triphenylphosphonium bromide 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23096-14
B23096-22
B23096-36
CAAAB23096-14EA
61.78
CAD
CAAAB23096-14
CAAAB23096-22
CAAAB23096-36
(Ethyl)triphenylphosphonium bromide 98+%
(Ethyl)triphenylphosphonium bromide
Purity: 98+%
CAS No.: 1530-32-1
Molecular Formula: C20H20BrP
Formula Weight: 371.26
Flash Point:
Formula:
C₂₀H₂₀BrP Melting Pt: 206…210 °C Flash Pt: >200 °C (392 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011838 CAS Number: 1530-32-1 EINECS: 216-223-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...