3-Chlorobenzophenone 98+%

Supplier: Thermo Scientific Chemicals
Warning

B23085-06 B23085-14 B23085-22
CAAAB23085-06EA 57.51 CAD
CAAAB23085-06 CAAAB23085-14 CAAAB23085-22
3-Chlorobenzophenone 98+%
3-Chlorobenzophenone

Purity: 98+% CAS No.: 1016-78-0 Molecular Formula: C13H9ClO Formula Weight: 216.67 Melting Point: 81-85° Beilstein Reference: 1869822

Formula: C₁₃H₉ClO
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00009816
CAS Number: 1016-78-0
EINECS: 213-809-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR