2,4-Dimethylphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B23076-03 B23076-06
CAAAB23076-03EA 109.18 CAD
CAAAB23076-03 CAAAB23076-06 CAAAB23076-14
2,4-Dimethylphenylboronic acid 97%
2,4-Dimethylphenylboronic acid

Purity: 97% CAS No.: 55499-44-0 Molecular Formula: C8H11BO2 Formula Weight: 149.99 Melting Point: 204-208° MDL No.: MFCD02683101

Formula: C₈H₁₁BO₂
MW: 149.98
Melting Pt: 204…208 °C
Storage Temperature: Ambient
MDL Number: MFCD02683101
CAS Number: 55499-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR