2-Thiopheneboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Thiophen-2-ylboronic acid, Thiophene-2-boronic acid, 2-Thienylboronic acid

B23071-03 B23071-06 B23071-14
CAAAB23071-03EA 44.08 CAD
CAAAB23071-03 CAAAB23071-06 CAAAB23071-14
2-Thiopheneboronic acid 98%
2-Thiopheneboronic acid

Purity: 98% CAS No.: 6165-68-0 Molecular Formula: C4H5BO2S Formula Weight: 127.96 Storage Temperature: 2°C to 8°C Melting Point: 128-131° Beilstein Reference: 112375 MDL No.: MFCD00151850

Formula: C₄H₅BO₂S
MW: 127.96 g/mol
Melting Pt: 128…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00151850
CAS Number: 6165-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR