3-(Dimethylamino)phenol 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23067-06
B23067-14
B23067-22
CAAAB23067-06EA
51.51
CAD
CAAAB23067-06
CAAAB23067-14
CAAAB23067-22
3-(Dimethylamino)phenol 97+%
3-(Dimethylamino)phenol
Purity: 97+%
CAS No.: 99-07-0
Molecular Formula: C8H11NO
Formula Weight: 137.18
Flash Point: 148°(298°F)
Odor: Pungent
Boiling Point: 267°
Beilstein Reference: 774631
MDL No.: MFCD00002264
Formula:
C₈H₁₁NO Boiling Pt: 267 °C Melting Pt: 80…84 °C Flash Pt: 148 °C(298 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002264 CAS Number: 99-07-0 EINECS: 202-727-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...